Anti-LRRK2 Rabbit Monoclonal Antibody [clone: EPR24565-56]
Supplier: Abcam
Certificates
About this item
Frequently Bought Together
CryoBoxes with hinged lid, for cryovials, 1 to 5 ml, TruCool®
Supplier: BIOCISION
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
Cell culture system, Corning® CELLine™ 1000
Supplier: Corning
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
Cryoboxes, B69/B50/B80/B99-Series
Supplier: GLW
Lab Consumables
Supplier: Copan
Serological pipettes, Stripette™
Supplier: Corning
VWR® Disposable Microcentrifuge Tubes
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Cryoboxes, 133×133 mm
Supplier: TENAK
VWR® Electroporation Cuvettes
Supplier: VWR Collection
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
Supplier: EPPENDORF
Screw caps for microcentrifuge tubes
Supplier: Thermo Fisher Scientific
OmniTop Sample Tubes® Exact Volume Sampling System (EVSS)
Supplier: Avantor Fluid Handling
Spinner flasks, disposable, Corning®
Supplier: Corning
Microcentrifuge tubes, 0,5/1,5/1,8 ml, Micrewtube®
Supplier: Simport Scientific
VWR®, Cryo Boxes
Supplier: VWR Collection