Anti-Jagged1 Rabbit Monoclonal Antibody [clone: EPR23647-75]
: Abcam
New Product
Rabbit monoclonal [EPR23647-75] to Jagged1.
Type: Primary
Antigen: Jagged1
Clonality: Monoclonal
Clone: EPR23647-75
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Frequently Bought Together