Anti-CD69 Rabbit Monoclonal Antibody [clone: EPR21814] (Alexa Fluor® 647)
Supplier: Abcam
Certificates
About this item
Alexa Fluor® 647 Rabbit monoclonal [EPR21814] to CD69.
Type: Primary
Antigen: CD69
Clonality: Monoclonal
Clone: EPR21814
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Frequently Bought Together