Anti-Pancreatic Polypeptide Rabbit Monoclonal Antibody [clone: EPR23320-10]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR23320-10] to Pancreatic Polypeptide.
Type: Primary
Antigen: Pancreatic Polypeptide
Clonality: Monoclonal
Clone: EPR23320-10
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Frequently Bought Together