Anti-galectin 9/Gal-9 Rabbit Monoclonal Antibody [clone: EPR23803-104]
Supplier: Abcam
Certificates
About this item
Rabbit monoclonal [EPR23803-104] to galectin 9/Gal-9 - BSA and Azide free (Capture).
Type: Primary
Antigen: galectin 9/Gal-9
Clonality: Monoclonal
Clone: EPR23803-104
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Frequently Bought Together