Anti-IgG2a Mouse Monoclonal Antibody [clone: MOPC-173]
Katalogové číslo ABCAAB18413-500
Dodavatel: Abcam
Specifikace
- Size:500 µg
- Pack type:Vial
Specifikace
O tomto produktu
Mouse IgG2a, Kappa Monoclonal [MOPC-173] -Isotype Control - ChIP Grade.
Type: Control
Antigen: IgG2a
Clonality: Monoclonal
Clone: MOPC-173
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human