Anti-VE Cadherin Rabbit Monoclonal Antibody [clone: EPR19106-67]
Supplier: Abcam
Certificates
About this item
Rabbit monoclonal [EPR19106-67] to VE Cadherin - BSA and Azide free (Detector).
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: VE Cadherin
Clonality: Monoclonal
Clone: EPR19106-67
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- ABCAAB242879-100
- ABCAAB242879-1
- Antigen symbol:
- VE Cadherin
- VE Cadherin
- Antigen name:
- CD144
- CD144
- Antigen synonyms:
- Cadherin-5|7B4 antigen|Vascular endothelial cadherin|VE-cadherin|CDH5
- Cadherin-5|7B4 antigen|Vascular endothelial cadherin|VE-cadherin|CDH5
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EPR19106-67
- EPR19106-67
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information.
- The exact immunogen used to generate this antibody is proprietary information.
- Form:
- Liquid
- Liquid
- Molecular weight:
- 87528 Da
- 87528 Da
- Purification:
- Protein A Affinity purification
- Protein A Affinity purification
- Storage temperature:
- Store at 4 °C.
- Store at 4 °C.
- Shipping temperature:
- 4 °C
- 4 °C
- Tested applications:
- sELISA
- sELISA
Specifications
Frequently Bought Together