Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Dengue Virus NS1 glycoprotein Mouse Monoclonal Antibody [clone: JE1.C10.H2.A9]
Anti-Dengue Virus NS1 glycoprotein Mouse Monoclonal Antibody [clone: JE1.C10.H2.A9]
  ABCAAB253231-100
 :  Abcam
undefined
Anti-Dengue Virus NS1 glycoprotein Mouse Monoclonal Antibody [clone: JE1.C10.H2.A9]
  ABCAAB253231-100
 :  Abcam

 

  • Seq:
    1286
  • Size:
    100 µg
  • Pack type:
    Vial

 

 

Mouse monoclonal [JE1.C10.H2.A9] to Dengue Virus NS1 glycoprotein.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Control
Antigen: IgG
Clonality: Monoclonal
Clone: PIP
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Armenian hamster
Isotype: IgG
Reactivity: