Anti-PD-L1 Rabbit Monoclonal Antibody [clone: 73-10] (Alexa Fluor® 555)
ABCAAB274896-100
: Abcam
- Size:100 µl
- Pack type:Vial
Alexa Fluor® 555 Rabbit monoclonal [73-10] to PD-L1.
Type: Primary
Antigen: PD-L1
Clonality: Monoclonal
Clone: 73-10
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human