Anti-BCMA Rabbit Monoclonal Antibody [clone: EPR28583-5]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR28583-5] to BCMA.
Type: Primary
Antigen: BCMA
Clonality: Monoclonal
Clone: EPR28583-5
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Frequently Bought Together