Anti-MUC2 Rabbit Monoclonal Antibody [clone: EPR23479-47]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR23479-47] to MUC2.
Type: Primary
Antigen: MUC2
Clonality: Monoclonal
Clone: EPR23479-47
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Frequently Bought Together