Order Entry
Czech Republic
ContactUsLinkComponent
Anti-IgG1 Mouse Antibody [clone: MOPC-21] (Alexa Fluor® 488)
Anti-IgG1 Mouse Antibody [clone: MOPC-21] (Alexa Fluor® 488)
Catalog # ABCAAB234075-100
Supplier:  Abcam
undefined
Anti-IgG1 Mouse Antibody [clone: MOPC-21] (Alexa Fluor® 488)
Catalog # ABCAAB234075-100
Supplier:  Abcam

Specifications

  • Seq:
    1286
  • Size:
    100 µg
  • Pack type:
    Vial

Specifications

About this item

AlexaFluor ® 488 Mouse IgG1 [MOPC-21] - Isotype Control.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Control
Antigen: IgG1
Clonality:
Clone: MOPC-21
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: