Anti-Flotillin 2/ESA Rabbit Monoclonal Antibody [clone: EPR26975-52]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR26975-52] to Flotillin 2/ESA.
Type: Primary
Antigen: Flotillin 2/ESA
Clonality: Monoclonal
Clone: EPR26975-52
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Frequently Bought Together