Anti-Androgen Receptor Rabbit Monoclonal Antibody [clone: EPR1535(2)]
Supplier: Abcam
Certificates
About this item
Rabbit monoclonal [EPR1535(2)] to Androgen Receptor - BSA and Azide free.
Type: Primary
Antigen: Androgen Receptor
Clonality: Monoclonal
Clone: EPR1535(2)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Frequently Bought Together