Anti-IL-1 beta Rabbit Monoclonal Antibody [clone: EPR16805-15]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR16805-15] to IL-1 beta.
Type: Primary
Antigen: IL-1 beta
Clonality: Monoclonal
Clone: EPR16805-15
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat
Frequently Bought Together