Anti-ICAM1 Rabbit Monoclonal Antibody [clone: EPR24639-3]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR24639-3] to ICAM1.
Type: Primary
Antigen: ICAM1
Clonality: Monoclonal
Clone: EPR24639-3
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Rat
Frequently Bought Together