Anti-Transferrin Receptor Mouse Monoclonal Antibody [clone: MEM-75] (Alexa Fluor® 700)
ABCAAB270667-100
: Abcam
- Seq:790
- Size:100 tests
- Pack type:Vial
Alexa Fluor® 700 Mouse monoclonal [MEM-75] to Transferrin Receptor.
: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Transferrin Receptor
Clonality: Monoclonal
Clone: MEM-75
Conjugation: Alexa Fluor® 700
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human