Anti-CD8 Mouse Monoclonal Antibody [clone: MEM-31] (PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®)
Catalog # ABCAAB157306-50
Supplier: Abcam
Specifications
- Rozměry protilátky:50 Tests
- Typ balení:Vial
Specifications
About this item
PerCP/Cy5.5® Mouse monoclonal [MEM-31] to CD8.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: CD8
Clonality: Monoclonal
Clone: MEM-31
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®
Epitope: ab157306 recognizes a conformationally-dependent epitope of CD8, a cell surface glycoprotein found on most cytotoxic T lymphocytes that mediates efficient cell-cell interactions within the immune system
Host: Mouse
Isotype: IgG2a
Reactivity: Human