Order Entry
Czech Republic
ContactUsLinkComponent
Anti-IGHG4 Mouse Antibody [clone: MOPC-21] (APC (Allophycocyanin)/Cy7®)
Anti-IGHG4 Mouse Antibody [clone: MOPC-21] (APC (Allophycocyanin)/Cy7®)
  ABCAAB239463-100
 :  Abcam
undefined
Anti-IGHG4 Mouse Antibody [clone: MOPC-21] (APC (Allophycocyanin)/Cy7®)
  ABCAAB239463-100
 :  Abcam

 

  • Seq:
    1286
  • Size:
    100 µg
  • Pack type:
    Vial

 

 

APC/Cy7® Mouse IgG1 kappa [MOPC-21] - Isotype Control.

 : All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Control
Antigen: IGHG4
Clonality:
Clone: MOPC-21
Conjugation: APC (Allophycocyanin)/Cy7®
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: