Anti-CDX-2 Mouse Monoclonal Antibody [clone: YN00475m]
Supplier: ANTIBODIES.COM
New Product
Certificates
About this item
Frequently Bought Together
Nalgene®, Biotainer® Bottles
Supplier: Thermo Fisher Scientific
VWR® Premium Line, Serological Pipettes
Supplier: VWR Collection
VWR®, Square Bottles, PET
Supplier: LAB3 OWN PRODUCTION
Petri dishes, Corning® Gosselin™
Supplier: Corning
Strips of 8 PCR tubes with attached caps, 0,2 ml, Amplitube™
Supplier: Simport Scientific
Petri dishes, square
Supplier: Greiner Bio-One
Multiwell cell culture plates, Falcon®
Supplier: Corning
Filter plates, 96-well, for multiplexing, AcroPrep™ Advance
Supplier: Cytiva (Formerly Pall Lab)
VWR®, Cryoboxes, without or with Dividers
Supplier: VWR Collection
Cap lock for microtube
Supplier: Greiner Bio-One
Spare Lid of Ice Pan 4L, colour option, RED, BLUE, GREEN, BLACK 1 * 1 items
Supplier: VWR Collection
CYTOONE 24-WELL PLATE, TC-TREATED, CLEA
Supplier: Life Systems Design AG
Multiple well cell culture plates, Costar®
Supplier: Corning
Screw cap trays for 2D barcoded storage tubes
Supplier: THERMO MATRIX TECHNOLOGIES
VIAL 50ML MLP W. FLIPTOP F. QUALITY DHIA 1 * 500 items
Supplier: Capitol Europe
Cryoboxes, 133×133 mm
Supplier: RATIOLAB
Detectable scoops, SteriPlast®
Supplier: BURKLE
BOTTLE PET 250ML SQUARE T/E GREEN CAP 1 * 200 items
Supplier: LAB3 OWN PRODUCTION