Specifications
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Mouse
- Target protein:Antithrombin III/ATIII
- Description:Mouse Antithrombin III/ATIII ELISA kit
- Sample type:Serum, plasma, tissue homogenates, and other biological fluids
- Cross reactivity:Mouse Antithrombin III/ATIII ELISA Kit exhibits high specificity and excellent specificity for the detection of mouse Antithrombin III/ATIII. No significant cross-reactivity or interference between Antithrombin III/ATIII and analogues was observed.
- Detection method:Colorimetric
- Time to Results:4 hours
- Shelf life:Store for 6 months at 4 °C
- Detection range:0,313 - 20 ng/ml
- Storage temperature:4 °C
- Sample volume:100 μl
- Sensitivity:0,188 ng/ml
- Regulatory status:RUO
- Tests per kit:96 tests
Specifications
About this item
Mouse Antithrombin III/ATIII ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Antithrombin III/ATIII in serum, plasma, tissue homogenates, and other biological fluids.
- Ready-to-use ELISA kit
- Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%
Mouse Antithrombin III/ATIII ELISA Kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse Antithrombin III/ATIII in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Antithrombin III/ATIII has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Antithrombin III/ATIII present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Antithrombin III/ATIII Antibody, which binds the captured Antithrombin III/ATIII present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Antithrombin III/ATIII captured in each well. The concentration of Antithrombin III/ATIII can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.