Human Recombinant IL37 (from E. coli)
Catalog # ANTIA317635-50
Supplier: Antibodies.com
New Product
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Seq:1202
- Storage conditions:Lyophilised: Store at −20...−80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Gene ID:UniprotID# Q9NZH6
- Reconstitution instructions:Reconstitute with distilled sterile water
- Carrier-free:N
- Protein synonyms:FIL1 zeta|FIL1Z|IL-1 zeta|IL-1H|IL-1H4|IL-1RP1|IL-1X|IL-37|IL1F7|IL1H4
- Protein/peptide name:IL37
- Purity:>95% by SDS-PAGE under reducing conditions
- Molecular weight:18,7 kDa
- Concentration:Reconstitution dependent
- Formulation:Lyophilised from a 0,2 µm filtered solution of 20 mM Phosphate Buffer, 150 mM NaCl, 2 mM DTT, pH 7,4
- Shipping temperature:Shipped on blue ice at 4 °C
- Tested applications:SDS-PAGE
Specifications
About this item
Partial length Human IL37, expressed in Escherichia coli.
Caution: This product is for Research Use Only. It is not for use in diagnostic or therapeutic procedures.