Order Entry
Czech Republic
ContactUsLinkComponent
Cat Recombinant IL-31 (from HEK293 cells)
Cat Recombinant IL-31 (from HEK293 cells)
Supplier:  Antibodies.com
New Product
Certificates
Cat Recombinant IL-31 (from HEK293 cells)
Supplier:  Antibodies.com
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Cat
  • Storage conditions:
    Lyophilised: Store at −20...−80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
  • Gene ID:
    UniprotID# XP_011286140.1
  • Reconstitution instructions:
    Reconstitute with distilled sterile water
  • Carrier-free:
    N
  • Protein synonyms:
    IL31|Interleukin-31
  • Protein/peptide name:
    IL-31
  • Purity:
    >85% by SDS-PAGE and coomassie blue staining
  • Molecular weight:
    The protein has a predicted molecular mass of 16,2 kDa after removal of the signal peptide. The apparent molecular mass of fIL31-His is approximately 15 - 25 kDa due to glycosylation.
  • Sequence:
    IL31 (Ala27-Gln159) + 6×His tag
  • Concentration:
    Reconstitution dependent
  • Formulation:
    Lyophilised from sterile Phosphate Buffered Saline, pH 7,4. Normally 5 - 8% trehalose is added as protectants before lyophilisation.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Tested applications:
    SDS-PAGE

Specifications

Frequently Bought Together