Order Entry
Czech Republic
ContactUsLinkComponent
Human Recombinant CD47 (from HEK293 cells)
Human Recombinant CD47 (from HEK293 cells)
Supplier:  Antibodies.com
New Product
Certificates
Human Recombinant CD47 (from HEK293 cells)
Supplier:  Antibodies.com
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    Lyophilised: store at −20...−80 °C. Reconstituted: aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
  • Gene ID:
    UniprotID# Q08722
  • Reconstitution instructions:
    Reconstitute with distilled sterile water.
  • Carrier-free:
    N
  • Protein synonyms:
    Antigenic surface determinant protein OA3|IAP|Integrin-associated protein|Leukocyte surface antigen CD47|MER6|Protein MER6
  • Protein/peptide name:
    CD47
  • Purity:
    >95% by SDS-PAGE and coomassie blue staining
  • Molecular weight:
    The protein has a predicted molecular mass of 60 kDa after removal of the signal peptide
  • Sequence:
    CD47 (Gln19-Gln141) (Val198-Ala207) (Ser257-Gly268) + mFc (Pro99-Lys330) + 6×His Tag
  • Concentration:
    Reconstitution dependent
  • Formulation:
    Lyophilised from sterile phosphate buffered Saline, pH 7,4, Normally 5 – 8% Trehalose is added as a protectant before lyophilisation
  • Shipping temperature:
    Shipped on blue ice at 4 °C.
  • Tested applications:
    ELISA, SDS-PAGE

Specifications

Frequently Bought Together