Order Entry
Czech Republic
ContactUsLinkComponent
Cynomolgus macaque Recombinant CD93 (from HEK293 cells)
Cynomolgus macaque Recombinant CD93 (from HEK293 cells)
Supplier:  Antibodies.com
New Product
Certificates
Cynomolgus macaque Recombinant CD93 (from HEK293 cells)
Supplier:  Antibodies.com
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Cynomolgus macaque
  • Storage conditions:
    Lyophilised: Store at −20...−80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
  • Gene ID:
    UniprotID# A0A2K5VH53
  • Reconstitution instructions:
    Reconstitute with distilled sterile water
  • Carrier-free:
    N
  • Protein synonyms:
    C1q/MBL/SPA receptor|C1qR|C1qR(p|C1QR1|C1qRp|CDw93|Complement component 1 q subcomponent receptor 1|Complement component C1q receptor|Matrix-remodeling-associated protein 4|MXRA4
  • Protein/peptide name:
    CD93
  • Purity:
    >85% by SDS-PAGE and coomassie blue staining
  • Molecular weight:
    The protein has a predicted molecular mass of 59,2 kDa after removal of the signal peptide. The apparent molecular mass of cCD93-His is approximately 70 - 130 kDa due to glycosylation.
  • Sequence:
    CD93 (Ala24-Leu581) + 6×His Tag
  • Concentration:
    Reconstitution dependent
  • Formulation:
    Lyophilised from sterile Phosphate Buffered Saline, pH 7,4. Normally 5 - 8% Trehalose is added as a protectant before lyophilisation.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Tested applications:
    SDS-PAGE

Specifications

Frequently Bought Together