Human Recombinant CD98 (from HEK293 Cells)
Dodavatel: Antibodies.com
New Product
Certifikáty
O tomto produktu
Partial length Human CD98, expressed in HEK293 cells.
Caution: This product is for esearch use only. It is not for use in diagnostic or therapeutic procedures.
Specifikace
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:Lyophilised: Store at −20...−80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Gene ID:UniprotID# P08195
- Reconstitution instructions:Reconstitute with distilled sterile water
- Carrier-free:N
- Protein synonyms:4F2 cell-surface antigen heavy chain|4F2 heavy chain antigen|4F2hc|Amino acid transporter heavy chain SLC3A2|Lymphocyte activation antigen 4F2 large subunit|MDU1|Solute carrier family 3 member 2
- Protein/peptide name:CD98
- Purity:>85% by SDS-PAGE and Coomassie blue staining
- Molecular weight:The protein has a predicted molecular mass of 47,7 kDa after removal of the signal peptide The apparent molecular mass of His-CD98 is approximately 55 - 100 kDa due to glycosylation
- Sequence:6×His Tag + CD98 (Arg206-Ala630)
- Concentration:Reconstitution dependent
- Formulation:Lyophilised from sterile phosphate buffered saline, pH 7,4. Normally 5 - 8% trehalose is added as a protectant before lyophilisation.
- Shipping temperature:Shipped on blue ice at 4 °C
- Tested applications:SDS-PAGE
Specifikace
Frequently Bought Together