Human Recombinant FGFR3b (from HEK293 Cells)
New Product
Partial length Human FGFR3b, expressed in HEK293 cells, suitable for WB, SDS-PAGE, Immunoassays, Functional Studies and is biologically active.
: This product is for Research Use Only. It is not for use in diagnostic or therapeutic procedures.
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:Upon delivery aliquot and store at −20 or −70 °C. Avoid freeze/thaw cycles.
- Endotoxin content:<0,1 ng/µg (1 IEU/µg), as measured by LAL method.
- Gene ID:UniprotID# P22607-2
- Carrier-free:N
- Protein/peptide name:FGFR3b
- Purity:>95% (by SDS-PAGE)
- Molecular weight:41,2 kDa (including tags).
- Sequence:ESLGTEQRVVGRAAEVPGPEPGQQEQLVFGSGDAVELSCPPPGGGPMGPTVWVKDGTGLVPSERVLVGPQRLQVLNASHEDSGAYSCRQRLTQRVLCHFSVRVTDAPSSGDDEDGEDEAEDTGVDTGAPYWTRPERMDKKLLAVPAANTVRFRCPAAGNPTPSISWLKNGREFRGEHRIGGIKLRHQQWSLVMESVVPSDRGNYTCVVENKFGSIRQTYTLDVLERSPHRPILQAGLPANQTAVLGSDVEFHCKVYSDAQPHIQWLKHVEVNGSKVGPDGTPYVTVLKSWISESVEADVRLRLANVSERDGGEYLCRATNFIGVAEKAFWLSVHGPRAAEEELVEADEAGSVYAGGSGHHHHHHHHHH
- Concentration:Lot specific
- Formulation:Supplied in Phosphate Buffered Saline with 20% Glycerol
- Shipping temperature:Shipped on blue ice at 4 °C
- Tested applications:WB, SDS-PAGE, Immunoassays, Functional Studies
Frequently Bought Together