Recombinant Human DLK-1 (from HEK293 cells)
Supplier: ANTIBODIES.COM
New Product
Certificates
About this item
Frequently Bought Together
CryoBoxes with hinged lid, for cryovials, 1 to 5 ml, TruCool®
Supplier: BIOCISION
Whatman™ Cellulose Blotting Papers, Grade GB003, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
VWR® Cryoboxes, Cardboard, 136×136 mm, with Full Labelling
Supplier: VWR Collection
Cell culture plates, 384-well, Small Volume™, CELLSTAR®
Supplier: Greiner Bio-One
Optical bottom plates, 96- and 384-well, Nunc™
Supplier: Thermo Fisher Scientific
Cellware, collagen Type I, Corning® BioCoat™
Supplier: Corning
Cell culture roller bottles, Corning®
Supplier: Corning
Storage tubes, 2D barcoded, Matrix™
Supplier: THERMO MATRIX TECHNOLOGIES
Serological pipettes
Supplier: GILSON
Strips of 8 PCR tubes with attached caps, 0,2 ml, Amplitube™
Supplier: Simport Scientific
Culture slides, Falcon®
Supplier: Corning
Corning® Gosselin™ Petri Dish, Aseptic
Supplier: Corning
Microcentrifuge tubes, 0,5/1,5/1,8 ml
Supplier: Simport Scientific