Recombinant Human PCDH18 (from HEK293 cells)
New Product
Frequently Bought Together
Serological pipettes, Stripette™
: Corning
VWR®, Petri Dish, PS
: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
: VWR Collection
Eppendorf® Protein LoBind® Microcentrifuge Tubes
: EPPENDORF
VWR® Standard Line Transfer Pipettes
: VWR Collection
Eppendorf® Safe-Lock Tubes, 0,5 ml - Microtubes
: EPPENDORF
Eppendorf Tubes® 3810X Microtubes 1,5 ml
: EPPENDORF
Serological pipettes
: Greiner Bio-One
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
: VWR Collection
VWR®, Transfer Pipettes, Graduated
: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF
VWR®, Cryoboxes, Cardboard, 133×133 mm
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection