Mouse Recombinant CD98 (from HEK293 cells)
Supplier: Antibodies.com
New Product
Certificates
About this item
Partial length Mouse CD98, expressed in HEK293 cells.
Caution: This product is for Research Use Only. It is not for use in diagnostic or therapeutic procedures.
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Storage conditions:Lyophilised: Store at −20...−80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Gene ID:UniprotID# P10852
- Reconstitution instructions:Reconstitute with distilled sterile water
- Carrier-free:N
- Protein synonyms:4F2 cell-surface antigen heavy chain|4F2 heavy chain antigen|4F2hc|Amino acid transporter heavy chain SLC3A2|Lymphocyte activation antigen 4F2 large subunit|MDU1|Solute carrier family 3 member 2
- Protein/peptide name:CD98
- Purity:>85% by SDS-PAGE and coomassie blue staining
- Molecular weight:The protein has a predicted molecular mass of 48,9 kDa after removal of the signal peptide. The apparent molecular mass of His-mCD98 is approximately 55 - 70 kDa due to glycosylation.
- Sequence:10×His tag + Mouse CD98 (Ala100-Ala526)
- Concentration:Reconstitution dependent
- Formulation:Lyophilised from sterile Phosphate Buffered Saline, pH 7,4. Normally 5 - 8% trehalose is added as protectants before lyophilisation.
- Shipping temperature:Shipped on blue ice at 4 °C
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together