Recombinant Cynomolgus macaque ErbB3/HER3 (from HEK293 cells)
Supplier: ANTIBODIES.COM
New Product
Certificates
About this item
Frequently Bought Together
96 MicroWell™ plates, Nunclon™∆
Supplier: Thermo Fisher Scientific
VWR®, Stoppers, PE
Supplier: VWR Collection
Inoculating loops and needles, disposable
Supplier: Greiner Bio-One
Colour coders for cryovials, CryoColorCode, Nunc™
Supplier: Thermo Fisher Scientific
Filter papers for healthcare applications, grade 470, Whatman™
Supplier: Whatman products (Cytiva)
Sterilin™ Samples Containers with Screw Caps, PS, 60 to 250 ml
Supplier: Sterilin
Ice pans and buckets, multi-purpose, Corning®
Supplier: Corning
Blotting transfer papers
Supplier: Ahlstrom-Munksjö
Peqlab, PCR Tubes
Supplier: VWR Peqlab
CRYOBOX IN PP 81 POS. H50MM RED 1 * 5 items
Supplier: RATIOLAB
Screw caps for T501 sample tubes
Supplier: Simport Scientific
Spinner flasks, disposable, Corning®
Supplier: Corning
Cryostorage boxes, KeepIT-100™, WHEATON®
Supplier: DWK Life Sciences
BOTTLE PET 250ML SQUARE T/E GREEN CAP 1 * 200 items
Supplier: LAB3 OWN PRODUCTION
OmniTop Sample Tubes® Exact Volume Sampling System (EVSS)
Supplier: Avantor Fluid Handling