Cynomolgus macaque Recombinant IgHE (from HEK293 Cells)
Supplier: Antibodies.com
New Product
Certificates
About this item
Partial length Cynomolgus macaque IgHE, expressed in HEK293 cells.
Caution: This product is for research use only. It is not for use in diagnostic or therapeutic procedures.
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Cynomolgus macaque
- Storage conditions:Lyophilised: Store at –20...–80 °C. Reconstituted: Aliquot and store at –80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Gene ID:UniprotID# A0A7N9DC98
- Reconstitution instructions:Reconstitute with distilled sterile water
- Carrier-free:N
- Protein synonyms:Ig epsilon chain C region|Ig epsilon chain C region ND|IGHE|Immunoglobulin heavy constant epsilon
- Protein/peptide name:IgHE
- Purity:>85% by SDS-PAGE and Coomassie blue staining
- Molecular weight:The protein has a predicted molecular mass of 48,3 kDa after removal of the signal peptide. The apparent molecular mass of cIgHE-His is approximately 55 - 70 kDa due to glycosylation.
- Sequence:IgHE (Ala1-Gly432) + 6×His Tag
- Concentration:Reconstitution dependent
- Formulation:Lyophilised from sterile phosphate buffered saline, pH 7,4. Normally 5 - 8% Trehalose is added as a protectant before lyophilisation.
- Shipping temperature:Shipped on blue ice at 4 °C
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together