Recombinant Human FOLR2 (from HEK293 cells)
Supplier: ANTIBODIES.COM
New Product
Certificates
About this item
Frequently Bought Together
Transfer pipettes
Supplier: RATIOLAB
CryoBoxes with hinged lid, for cryovials, 1 to 5 ml, TruCool®
Supplier: BIOCISION
VWR® Media Bottles, Square, PETG
Supplier: VWR Collection
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
Measuring scoops
Supplier: VITLAB
VWR®, Cell Culture Flasks, Non Treated or Treated
Supplier: VWR Collection
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
Supplier: EPPENDORF
VWR®, Petri Dish, PS
Supplier: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
J.T.Baker®, Plates, Square Well
Supplier: Avantor
Round-bottom polystyrene tubes, disposable, Falcon®
Supplier: Corning
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
Deep well plates, 24-well, Axygen®
Supplier: Corning
VWR®, Cryo coder cap inserts
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Spreaders, L-shaped, Disposable
Supplier: VWR Collection
Spinner flasks, disposable, Corning®
Supplier: Corning