Human Recombinant ULBP2 (from HEK293 cells)
Supplier: Antibodies.com
New Product
Certificates
About this item
Partial length Human ULBP2, expressed in HEK293 cells.
Caution: This product is for Research Use Only. It is not for use in diagnostic or therapeutic procedures.
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:Lyophilised: Store at −20...−80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Gene ID:UniprotID# Q9BZM5
- Reconstitution instructions:Reconstitute with distilled sterile water
- Carrier-free:N
- Protein synonyms:ALCAN-alpha|N2DL-2|N2DL2|NKG2D ligand 2|NKG2DL2|RAET1H|Retinoic acid early transcript 1H|UL16-binding protein 2
- Protein/peptide name:ULBP2
- Purity:>85% by SDS-PAGE and coomassie blue staining
- Molecular weight:The protein has a predicted molecular mass of 22,5 kDa after removal of the signal peptide. The apparent molecular mass of ULBP2-His is approximately 15 - 35 kDa due to glycosylation.
- Sequence:ULBP2 (Gly26-Ser216) + 6×His Tag
- Concentration:Reconstitution dependent
- Formulation:Lyophilised from sterile Phosphate Buffered Saline, pH 7,4. Normally 5 - 8% Trehalose is added as a protectant before lyophilisation.
- Shipping temperature:Shipped on blue ice at 4 °C
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together