
Ethanol denatured ≥90% (denatured with 5% isopropanol and 5% methanol)
Catalog # VWRS83898.320
Supplier: VWR Collection
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
Specifications
About this item
:
:46.07 g/mol
Storage Temperature:Ambient
EINECS:200-578-6
UN:1170
ADR:3,II
:GHS02
CAS Number:64-17-5
Formula:H₃CCH₂OH
MDL Number:MFCD00003568