Ethyl-2-oxocyclopentanecarboxylate ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:611-10-9
:
Formula:C₈H₁₂O₃
MDL Number:MFCD00001412
Boiling Pt:225…228 °C
Density:1.079
Flash Pt:77 °C (170 °F)
:156.18 g/mol
Storage Temperature:Ambient
EINECS:210-253-0
Related Information
Frequently Bought Together