Order Entry
Czech Republic
ContactUsLinkComponent
 
 
 
Methyl-3-(bromomethyl)benzoate 95%
 
undefined
 
 
Methyl-3-(bromomethyl)benzoate 95%

 

  • Appearance:
    White Crystalline powder or crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    41 to 47 °C
  • GC:
    ≥94 %

 

 

Frequently Bought Together