Order Entry
Czech Republic
ContactUsLinkComponent
Lithiumbis(trimethylsilyl)amide 20% ∼1.06 M in THF/ethylbenzene
Flammable
Corrosive
Health
Irritant
Lithiumbis(trimethylsilyl)amide 20% ∼1.06 M in THF/ethylbenzene
Catalog # L15012.AE
CAS Number:  
Flammable
Corrosive
Health
Irritant
Lithiumbis(trimethylsilyl)amide 20% ∼1.06 M in THF/ethylbenzene
Catalog # L15012.AE
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

CAS Number:4039-32-1
 :
Formula:C₆H₁₈LiNSi₂
MDL Number:MFCD00008261
 :167.33 g/mol
Density:0.891
Flash Pt:–17 °C (1 °F)
Storage Temperature:Ambient
EINECS:223-725-6
UN:2924
ADR:3,II
 :GHS02
 :GHS05
 :GHS08
 :GHS07