Order Entry
Czech Republic
ContactUsLinkComponent
EEDQ (ethyl-N-(2-ethoxy-1,2-dihydroquinoline)carboxylate), Sigma-Aldrich®
EEDQ (ethyl-N-(2-ethoxy-1,2-dihydroquinoline)carboxylate), Sigma-Aldrich®
Catalog # SIALE103-1G
Supplier:  Merck
CAS Number:  
EEDQ (ethyl-N-(2-ethoxy-1,2-dihydroquinoline)carboxylate), Sigma-Aldrich®
Catalog # SIALE103-1G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :M-Clarity™ quality level = MQ100
 :247.29 g/mol
EINECS:240-418-2
CAS Number:16357-59-8
Formula:C₁₄H₁₇NO₃
MDL Number:MFCD00006703