Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Fluorene-[D10] 200 µg/ml in isooctane (98% D), CIL
Fluorene-[D10] 200 µg/ml in isooctane (98% D), CIL
Fluorene-[D10] 200 µg/ml in isooctane (98% D), CIL
Catalog # PROOCILDLM11231.2
Supplier:
LGC Standards PROMOCHEM
CAS Number:
Expand image
Fluorene-[D10] 200 µg/ml in isooctane (98% D), CIL
Catalog # PROOCILDLM11231.2
Supplier:
LGC Standards PROMOCHEM
CAS Number:
Specifications
Pack type:
Glass bottle
Pk:
1,2 mL
Description:
Fluorene-[D10] 200 µg/ml in isooctane (98% D)
Specifications
About this item
Product Details
Product Details