Order Entry
Czech Republic
ContactUsLinkComponent
Nicotinoyl chloride hydrochloride, Sigma-Aldrich®
Nicotinoyl chloride hydrochloride, Sigma-Aldrich®
Catalog # SIAL213381-25G
Supplier:  Merck
CAS Number:  
Nicotinoyl chloride hydrochloride, Sigma-Aldrich®
Catalog # SIAL213381-25G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :178.02 g/mol
ADR:8,II
 :M-Clarity™ quality level = MQ100
EINECS:243-648-1
UN:3261
CAS Number:20260-53-1
Formula:C₆H₅Cl₂NO
MDL Number:MFCD00012820