Order Entry
Czech Republic
ContactUsLinkComponent
Nicotinoyl chloride hydrochloride 97%
Corrosive
Nicotinoyl chloride hydrochloride 97%
Certificates
undefined
Corrosive
Nicotinoyl chloride hydrochloride 97%

Specification Test Results

  • Appearance:
    White to yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    151 to 156 °C
  • Titration RCOCl:
    96.0 to 104.0 %

Specification Test Results

Related Information

Frequently Bought Together