3-(4-Methoxyphenoxy)benzaldehyde, Sigma-Aldrich®
Catalog # SIAL195898-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
EINECS:263-527-7
CAS Number:62373-80-2
Formula:C₁₄H₁₂O₃
MDL Number:MFCD00003358