Order Entry
Czech Republic
ContactUsLinkComponent
(4-(Trifluoromethyl)phenyl)methanesulphonyl chloride ≥97%
Corrosive
(4-(Trifluoromethyl)phenyl)methanesulphonyl chloride ≥97%
Catalog # H26395.03
CAS Number:  
Corrosive
(4-(Trifluoromethyl)phenyl)methanesulphonyl chloride ≥97%
Catalog # H26395.03
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

About this item

CAS Number:163295-75-8
 :
Formula:C₈H₆ClF₃O₂S
MDL Number:MFCD01631932
 :258.65 g/mol
Melting Pt:103…105 °C
Storage Temperature:Ambient
EINECS:None
UN:3261
ADR:8,II
 :GHS05