Order Entry
Czech Republic
ContactUsLinkComponent
Acetaldehyde-[¹³C₂] (99% ¹³C ), Sigma-Aldrich®
Flammable
Health
Irritant
Acetaldehyde-[¹³C₂] (99% ¹³C ), Sigma-Aldrich®
Catalog # SIAL531227-1G
Supplier:  Merck
CAS Number:  
Flammable
Health
Irritant
Acetaldehyde-[¹³C₂] (99% ¹³C ), Sigma-Aldrich®
Catalog # SIAL531227-1G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
ADR:3,I
 :M-Clarity™ quality level = MQ200
UN:1089
 :GHS02
 :GHS08
 :GHS07
Storage Temperature:Refrigerator
CAS Number:1632-98-0
Formula:C₂H₄O
MDL Number:MFCD00143529