2-(Trimethylsilyloxy)-1,3-cyclohexadiene, Sigma-Aldrich®
Catalog # SIAL423424-5ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 mL
- Seq:2210
Specifications
About this item
:
:M-Clarity™ quality level = MQ100
CAS Number:54781-19-0
Formula:C₉H₁₆OSi
MDL Number:MFCD00274202