

Calcium sulphate, Drierite™ 3 mm (8 mesh) for drying, with indicator (cobalt (II) chloride)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7778-18-9
:
Formula:CaO₄S
MDL Number:MFCD00010912
:136.14 g/mol
Merck Index:14,1706
Storage Temperature:Ambient
EINECS:231-900-3
UN:3077
ADR:9,III
:GHS08
:GHS07
Related Information
Frequently Bought Together