Supelco® PESTANAL® Organic Reference Standard, Allyl Isothiocyanate
Catalog # 36682-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Standards:PESTANAL®, Analytical standard
- CAS No.:57-06-7
- Description:Allyl isothiocyanate
- Storage conditions:2...30 °C
- Hazard class description:H226 - H302 - H310 + H330 - H314 - H317 - H410
- Shipping conditions:STD
- MDL No.:MFCD00004822
- M-Clarity quality level:MQ100
- Synonym(s):AITC; Oil of mustard
- Melting point:–80 °C (lit.)
- Formula:CH₂=CHCH₂NCS
- Molecular weight:99,15
- Density:1,013 g/ml at 25 °C (lit.)
- Flash point:45 °C - closed cup
- Boiling point:150 °C (lit.)
Specifications
About this item
- GHS signal: GHS02, GHS06, GHS08, GHS09
Caution: P210 - P273 - P280 - P303 + P361 + P353 - P304 + P340 + P310 - P305 + P351 + P338