Order Entry
Czech Republic
ContactUsLinkComponent
Gallipots, graduated, Azlon®
Gallipots, graduated, Azlon®
Catalog # 216-1021
Supplier:  SciLabware
undefined
Gallipots, graduated, Azlon®
Catalog # 216-1021
Supplier:  SciLabware

Specifications

  • Version:
    non sterile
  • Capacity:
    60 ml
  • Pk:
    1.000

Specifications

About this item

PP, medical grade.

  • Moulded graduations to capacity
  • Autoclavable to 121 °C
  • Low form

Suitable for sampling, short term storage, micro-analysis or many other applications.