ContactUsLinkComponent
Refrigerators, integrated built-in combinations
Refrigerators, integrated built-in combinations
Catalog # LIEBICUS2913
Supplier:  Liebherr
Refrigerators, integrated built-in combinations
Catalog # LIEBICUS2913
Supplier:  Liebherr
Specifications
  • W×D×H ext.:
    1590×570×1574 mm
  • Capacity:
    247 l
  • Description:
    Built-in refrigerator with integrated freezer, ICUS 2913 Comfort
  • Model:
    ICUS 2913 Comfort
  • Cat. No.:
    LIEBICUS2913
  • Alarm:
    No alarm
  • Defrost:
    automatic (fridge)
    manual (freezer)
  • Display:
    LED
  • Door type:
    1 solid sliding door
  • Fans:
    no fan
  • Shelves/drawers:
    4 shelves (refrigerator), 2 drawers (freezer), 2 baskets
  • Energy class:
    A+
  • Pk:
    1
Specifications
About this item
These integrated built-in refrigerators are designed to maintain the temperature and an ideal air humidity.
  • Accurate temperature controller
  • Duocooling with two completely separate cooling circuits
  • Prevents odour transfer
  • Protects food from dryness